OM-ICTF13XB
OMICRON™
OMICRON™ PTFE (Hydrophilic) Syringe Filters, Non-Sterile, HPLC Certified
0.45 Micron Pore, 13mm, Non-Sterile, 100/Pk
Synonym(s):
Chromatography Syringe Filter, PTFE, Hydrophilic
About This Item
Recommended Products
manufacturer/tradename
Omicron Scientific
General description
Media: PTFE (Polytetrafluoro ethylene)
PTFE (Hydrohilic) Syringe Filters provide cleaner sample extracts by removing interfering materials and fine particles. They are optically clear when wet with water. They are versatile filters for use with aqueous and aggressive organic solvent based solutions and are especially ideal for HPLC sample preparation. These filters are used for filtration of highly basic or hot solutions, without need of pre-wetting the membrane. They can also filter aqueous solutions without pre-wetting.
Technical Specifications- Filtration Area: 0.8cm² for 13mm, 3.9cm² for 25mm
- Bubble Point: ≥20psi for 0.2µm & ≥14 psi for 0.45µm
- Water Flow: 15ml/min & 32mlmin for 13mm, 42ml/min & 115ml/min for 25mm (@ 10psi)
- Low extractable
- No need for pre-wetting
- High flow rates with minimal aqueous extractables
- Low surface friction co-efficient: 0.1
- High pH resistance
- HPLC sample preparation
- Routine QA/QC analysis
- Removal of protein precipitates
- Dissolution testing
- Venting applications & Degassing solvents
(Closely similar to 6774-1304)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.