Key Documents
OM-SFNY25XB
OMICRON™
OMICRON™ Nylon Syringe Filters, Non-Sterile, HPLC Certified
0.45 Micron Pore, 25mm, Non-Sterile, 100/Pk
Synonym(s):
Chromatography Syringe Filter, Nylon Syringe Filters
About This Item
PDF Icon
Shipped & Sold By Omicron Scientific
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Omicron Scientific
General description
Product Description
OMICRON™ Nylon Syringe Filters are manufactured with hydrophilic Nylon membrane which removes microorganisms, particles, precipitates, and undissolved solid particles for aqueous or organic solutions. These single use products consist of a nylon membrane filter sealed in PP (Polypropylene) housing and are manufactured using a unique impregnate process that eliminates cracking, tearing, breaking and distortion during use. OMICRON™ Nylon Syringe Filters have low extractable which assure them ideal for solvent filtration and sample preparation for analytical applications.
Key Features
  • Naturally hydrophilic
  • Very low sample absorption
  • Minimal hand pressure
  • HPLC certified for low UV extractables
Key Applications
  • Sample filtration of HPLC & LC-MS Analysis
  • Analytical Sample preparation and Clarification
  • Filtration of aqueous solutions
  • Suitable for method development processes
  • Sterile filtration of biological solutions and buffers
(Similar to 6753-2504)
Legal Information
OMICRON™ is a trademark of Omicron Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)