Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17189

Millipore

Park and Sanders Enrichment Broth (Base)

BioChemika, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

composition

casein enzymic hydrolysate, 10 g/L
dextrose, 1 g/L
monohydrogen sodium sulfite, 0.1 g/L
peptic digest of animal tissue, 10 g/L
sodium chloride, 5 g/L
sodium citrate, 1 g/L
sodium pyruvate, 0.25 g/L
yeast extract, 2 g/L

final pH

7.0±0.2 (25 °C)

application(s)

microbiology

Application

For selective enumeration of thermotolerant Campylobacter species from foods.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service