Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53275

Millipore

Hippurate Broth

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

shelf life

limited shelf life, expiry date on the label

Quality Level

composition

sodium hippurate

application(s)

food and beverages
microbiology

suitability

nonselective and differential for Campylobacter spp.

Application

For the demonstration of hippurate hydrolysis (Campylobacter jejuni).

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Harvey
Journal of clinical microbiology, 11(4), 435-437 (1980-04-01)
An additional method for differentiating between Campylobacter fetus subsp. jejuni and C. fetus subsp. intestinalis is reported. Strains of C. fetus subsp. jejuni (18/20) were shown to hydrolyze hippurate in the 2-h rapid test, whereas strains of C. fetus subsp.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service