Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV38672

Sigma-Aldrich

Anti-TSFM (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Ts translation elongation factor, mitochondrial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

human, rabbit, horse, bovine, guinea pig, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TSFM(10102)

Immunogen

Synthetic peptide directed towards the C terminal region of human TSFM

Biochem/physiol Actions

Ts translation elongation factor, mitochondrial (TSFM) is a guanine nucleotide exchange factor for EFTu during the elongation step of protein translation in mitochondria. Mutations in TSFM gene results in combined oxidative phosphorylation deficiency-3 syndrome.

Sequence

Synthetic peptide located within the following region: VVGMAPLSVGSLDDEPGGEAETKMLSQPYLLDPSITLGQYVQPQGVSVVD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonia Emperador et al.
European journal of human genetics : EJHG, 25(1), 153-156 (2016-09-30)
Oxidative phosphorylation dysfunction has been found in many different disorders. This biochemical pathway depends on mitochondrial protein synthesis. Thus, mutations in components of the mitochondrial translation system can be responsible for some of these pathologies. We identified a new homozygous
Assignment of the mitochondrial translation elongation factor Ts gene (TSFM) to human chromosome 12 bands q13-->q14 by in situ hybridization and with somatic cell hybrids.
J L Vernon et al.
Cytogenetics and cell genetics, 89(3-4), 145-146 (2000-08-31)
Jan A M Smeitink et al.
American journal of human genetics, 79(5), 869-877 (2006-10-13)
The 13 polypeptides encoded in mitochondrial DNA (mtDNA) are synthesized in the mitochondrial matrix on a dedicated protein-translation apparatus that resembles that found in prokaryotes. Here, we have investigated the genetic basis for a mitochondrial protein-synthesis defect associated with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service