Key Documents
BMSNS1020
Benchmark SmartDrop XF Nano Spectrophotometer
new
AC/DC input 115 V, US 2-pin plug
About This Item
UNSPSC Code:
41121800
manufacturer/tradename
Benchmark Scientific NS1020
General description
Accuris DNA Quantification Kits are designed for accurate fluorescent quantification of dsDNA using instruments such as the Qubit, SmartDrop XF, and microplate readers. The reagents are highly selective for double-stranded DNA (dsDNA); quantification results will not be skewed by single-stranded DNA (ssDNA), RNA, protein, or nucleotides in a sample.  Included with each kit are a concentrated fluorescent assay reagent, dilution buffer, and ready-to-use DNA standards.  The assay reagent is diluted using the buffer provided, sample is added (1 µL to 20 µL), and the concentration determined using an appropriate fluorometer instrument that utilizes blue excitation (460 ± 20 nm) and measures green fluorescence (525-570 nm).High Sensitivity dsDNA Assay KitThe Accuris High Sensitivity Kit provides an accurate method to quantify low concentration dsDNA samples.  The assay kit is designed to be accurate for initial DNA sample concentrations of 0.005–120 ng/µl, providing a detection range of 0.1–120 ng.Broad Range dsDNA Assay KitThe Accuris Broad Range Kit provides an accurate method to quantify dsDNA samples that have a broad concentration range starting at 0.2 ng/µL. The assay kit is designed to be accurate for initial DNA sample concentrations of 0.2 to 2,000 ng/µL, providing a detection range of 4−2,000 ng.
Application
Spectrophotometry, Colorimetry, OD600, UV-Vis Spectral Scanning, fluorimetry
Features and Benefits
  • Quick and accurate results from a sample droplet
  • Touch screen control panel, no PC or software required
  • Built-in printer and cuvette port for OD600 readings
  • XF model includes fluorometer for sensitive DNA
  • quantification
Choose from one of the most recent versions:
Certificates of Analysis (COA)