Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DUO87011

Sigma-Aldrich

Duolink® PLA® Probe Mouse Anti-PhosSer PLUS from mouse

Protein Phosphorylation Detection by Proximity Ligation Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody (Protein G-Sepharose)

antibody product type

primary antibodies

clone

4A4, monoclonal

product line

Duolink®

species reactivity

all

technique(s)

proximity ligation assay: suitable

suitability

suitable for brightfield
suitable for fluorescence

shipped in

wet ice

storage temp.

2-8°C

General description

Anti-Phosphoserine antibody, clone 4A4 with the Duolink® PLA® PLUS oligonucleotide. This antibody detects serine phosphorylated proteins in all species. Protein target specificity can be obtained by pairing with an antibody specific to a protein of interest in a Duolink® PLA® experiment. This product must be used in combination with a MINUS PLA probe of a different species or another directly-conjugated primary antibody with a MINUS PLA oligo. The Anti-mouse PLA probe PLUS (DUO92001) is not necessary when using this product.

Specificity

This directly-conjugated primary antibody PLA probe recognizes serine-phosphorylated proteins from all species.

Application

Duolink® proximity ligation assay(PLA®) allows for endogenous detection of protein interactions, post translational modifications, and protein expression levels at the single molecule level in fixed cells and tissue samples.

This product can be applied to both the Duolink® In Situ Fluorescence Protocol and the Duolink® In Situ Brightfield Protocol depending on the detection reagents used.

Visit our Duolink® PLA Resource Center for information on how to run a Duolink® experiment, applications, troubleshooting, and more.

To perform a complete Duolink® PLA in situ experiment you will need two primary antibodies (PLA, IHC, ICC or IF validated) that recognize two target epitopes. Other necessary reagents include a pair of PLA probes from different species (one PLUS and one MINUS), detection reagents, wash buffers, and mounting medium. Note that the primary antibodies must come from the same species as the Duolink® PLA probes. Analysis is carried out using standard immunofluorescence assay equipment.HRP is also available for brightfield detection.

Application Note
Protein phosphorylation is considered as one of the key steps in signal transduction and regulation of enzymatic activity. Anti-phosSer antibodies are commonly used in western blots after the targeted proteins have been immunoprecipitated to measure the Serine phosphorylation of the protein of interest. This process is time-consuming and cannot provide subcellular localization.

Anti-phosSer PLA probes are used to detect the phosphorylation of serine residues in proteins. Protein target specificity can be obtained by pairing with an antibody specific to a protein of interest in a Duolink® PLA® experiment. This product is particularly useful when site-specific anti-phosphorylation antibodies are not available. A MINUS PLA probe of a different species or another directly-conjugated primary antibody with a MINUS PLA oligo must be used in combination with this product. The Anti-mouse PLA probe PLUS (DUO92001) is not necessary when using this product. See our Product Selection Guide for more information.

Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects

Features and Benefits

  • No overexpression or genetic manipulation required
  • High specificity (fewer false positives)
  • Single molecule sensitivity due to rolling circle amplification
  • Relative quantification possible
  • No special equipment needed
  • Quicker and simpler than FRET
  • Increased accuracy compared to co-IP
  • Publication-ready results

Legal Information

Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service