Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU045171

Sigma-Aldrich

MISSION® esiRNA

targeting human RASGRP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGGGTCCAGTTGATGGTTCTTAGCAAACCAACCCCCCAGCAAAGGGCAGAAGTCATCACAAAGTTTATCAATGTTGCAAAGAAGCTCCTTCAGCTCAAAAATTTTAACACCCTGATGGCAGTGGTGGGAGGCCTCAGTCATAGTTCCATTTCACGCCTCAAAGAGACCCATTCTCATCTTTCTTCAGAAGTTACAAAGAACTGGAATGAAATGACAGAGTTGGTCTCCTCCAACGGCAATTACTGCAATTACCGCAAGGCCTTTGCCGACTGCGATGGCTTCAAAATCCCCATCCTTGGAGTACACTTGAAAGACTTGATAGCTGTCCATGTCATTTTCCCAGACTGGACAGAGGAGAACAAAGTGAACATTGTGAAAATGCACCAGCTCTCCGTTACCCTGAG

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang-Jie An et al.
American journal of translational research, 11(3), 1800-1809 (2019-04-12)
This study examined the relationship between the expression of Ras guanyl nucleotide-releasing protein 3 (RasGRP3) and disease activity in systemic lupus erythematosus (SLE) and explored the possible mechanisms in MRL/lpr mice. We detected the expression of RasGRP3 in peripheral blood
Ge Hu et al.
Oncology reports, 38(5), 2975-2984 (2017-10-20)
In the process of enlarging of tumors, the dissolving tissue structures and remodeling endothelial cells for restoring gas exchange and nutritional support, further facilitate tumor cell invasion and metastasis. Activation of Ras plays a critical role in the development of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service