Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU104911

Sigma-Aldrich

MISSION® esiRNA

targeting human PDZK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCTATGAGAAGGTGGTGGATAGAATCCAGAGCAGTGGGAAGAATGTCACACTTCTAGTCTGTGGAAAGAAGGCCTATGATTATTTCCAAGCTAAGAAAATCCCTATTGTTTCCTCCCTGGCTGATCCACTTGACACCCCTCCAGATTCTAAAGAAGGAATAGTGGTGGAGTCAAACCATGACTCGCACATGGCAAAAGAACGGGCCCACAGTACAGCCTCACATTCTTCTTCCAATTCTGAAGATACAGAGATGTGATGAAAACAAGTAATAGCTTTGGCTGTTTATTTGATAGCTGTTTCTGGGTATTTAATAGGAATCCTTTCTCAAGGAATGAGTTGTGACCTGTTTACTGTCTCTTTAGAAGAAAAACTCCACTGGAAACCATTCACCATGTGTGATTGTCTTCTGTTATCATTTGTCTTACAGGCGGCTATTGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Shao et al.
Oncology letters, 15(5), 6655-6665 (2018-04-05)
Patients with non-small-cell lung cancer (NSCLC) with sensitive epidermal growth factor receptor (EGFR) mutations generally react well to tyrosine kinase inhibitors (TKIs). However acquired resistance eventually occurs. Several mechanisms contribute to the resistance including T790M mutation, c-Met amplification and PIK3CA
Chunjuan Zhao et al.
Cancer letters, 453, 107-121 (2019-04-02)
Phosphorylation of PTEN plays an important role in carcinogenesis and progression of gastric cancer. However, the underlying mechanism of PTEN phosphorylation regulation remains largely elusive. In the present study, PDZK1 was identified as a novel binding protein of PTEN by
Ben J Wu et al.
International journal of cardiology, 199, 204-212 (2015-07-24)
High density lipoprotein (HDL) infusions increase new blood vessel formation (angiogenesis) in rodents with ischemic injury. This study asks if increasing HDL levels by inhibiting cholesteryl ester transfer protein (CETP) activity increases angiogenesis in New Zealand White (NZW) rabbits with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service