Key Documents
EPS44I042001
Eppendorf® Innova S44i Static Incubation Shelf
for use with Innova® S44i
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® S44I042001
compatibility
for use with Innova® S44i
Related Categories
General description
Static incubation shelf, Innova S44i.
Features and Benefits
  • Cannot be used with photosynthetic light bank installed
  • Note reduction in maximum flask size to 1 L Erlenmeyer or 3 L Fernbach
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
Choose from one of the most recent versions:
Certificates of Analysis (COA)