Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17372411

HiTrap® Capto Core 400

pack of 5 × 1 mL, Cytiva 17372411

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.21

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 17372411

General description

Novel core bead technology and multimodal ligand give Capto Core 400 dual functionality, size exclusion and binding chromatography, in one chromatography resin.

  • High capacity and productivity in flowthrough mode.
  • Prepacked HiTrap columns for easy screening and convenient process development, as well as small scale purification.
  • Wide operational window of pH and conductivity.


Capto Core 400 is an excellent alternative to size-exclusion chromatography (SEC) resins that are typically employed in the final stages of virus purification in vaccine manufacture.

Legal Information

Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

53.6 °F

flash_point_c

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service