Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PLA0202

Sigma-Aldrich

Rabbit anti-ATF2 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

Activating transcription factor 2, CRE-BP1, CREB-2, CREB2, HB16, TREB7, activating transcription factor 2 splice variant ATF2-var2, cAMP response element-binding protein CRE-BP1, cAMP responsive element binding protein 2, cAMP-dependent transcription factor ATF-2, cAMP-responsive element-binding protein 2, cyclic AMP-responsive element-binding protein 2, formerly, histone acetyltransferase ATF2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

AAY17203.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... ATF2(1386)

Immunogen

The epitope recognized by PLA0202 maps to a region between residue 437 and 487 of human activating transcription factor 2 using the numbering given in entry AAY17203.1 (GeneID 1386).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Activating transcription factor 2 (ATF2) is a leucine-zipper transcription factor that binds to the cAMP-responsive element (CRE). ATF2 is involved in regulating the expression of a variety of proteins that are important to oncogenesis. It is a substrate of the JNK/p38 and ATM/ATR protein kinases which regulate its transactivating activity in response to environmental signals.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service