R8157
RSK4, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
RPS6KA6
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
51111800
NACRES:
NA.32
Recommended Products
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
459-621 nmol/min·mg
mol wt
~112 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... RPS6KA6(27330)
Biochem/physiol Actions
RSK4 is a member of the 90-kDa ribosomal S6 kinase family which are important components of growth factor mediated stimulation of cellular proliferation, survival and differentiation. RSK4 is activated via coordinated phosphorylation by ERK and 3-phosphoinositide-dependent protein kinase-1 (PDK1). RSK4 is often associated with contiguous gene syndromes consisting of X-linked deafness type 3 (DFN3), mental retardation (MRX) and choroideremia (CHM). RSK4 is most abundantly expressed in the brain and kidney.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bettina A Dümmler et al.
The Journal of biological chemistry, 280(14), 13304-13314 (2005-01-06)
The 90-kDa ribosomal S6 kinases (RSK1-3) are important mediators of growth factor stimulation of cellular proliferation, survival, and differentiation and are activated via coordinated phosphorylation by ERK and 3-phosphoinositide-dependent protein kinase-1 (PDK1). Here we performed the functional characterization of a
H G Yntema et al.
Genomics, 62(3), 332-343 (2000-01-25)
Large deletions in Xq21 often are associated with contiguous gene syndromes consisting of X-linked deafness type 3 (DFN3), mental retardation (MRX), and choroideremia (CHM). The identification of deletions associated with classic CHM or DFN3 facilitated the positional cloning of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service