Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1305775

Sigma-Aldrich

ANTI-FEZF2 (CENTER) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

FEZF2, FEZL, Fez family zinc finger protein 2, Forebrain embryonic zinc finger-like protein 2, ZNF312

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

48811 Da

species reactivity

human

technique(s)

western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FEZF2(55079)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anupamaa Seshadri et al.
Critical care medicine, 45(9), 1523-1530 (2017-07-04)
Trauma induces a complex immune response that requires a systems biology research approach. Here, we used a novel technology, mass cytometry by time-of-flight, to comprehensively characterize the multicellular response to trauma. Peripheral blood mononuclear cells samples were stained with a
Emilie J Cosway et al.
The Journal of experimental medicine, 214(11), 3183-3195 (2017-08-24)
During αβT cell development, the thymus medulla represents an essential microenvironment for T cell tolerance. This functional specialization is attributed to its typical organized topology consisting of a branching structure that contains medullary thymic epithelial cell (mTEC) networks to support
D Hanidziar et al.
Scientific reports, 10(1), 4677-4677 (2020-03-15)
Prolonged exposure to hyperoxia has deleterious effects on the lung, provoking both inflammation and alveolar injury. The elements of hyperoxic injury, which result in high rates of lethality in experimental models, are thought to include multicellular immune responses. To characterize
Lindsey E Padgett et al.
Arteriosclerosis, thrombosis, and vascular biology, 40(12), 2845-2859 (2020-10-16)
Cardiovascular disease (CVD) remains a significant global health concern with a high degree of mortality. While CD4+ T cells have been extensively studied in CVD, the importance of CD8+ T cells in this disease, despite their abundance and increased activation
Akiko Matsuda et al.
Journal of applied toxicology : JAT, 40(5), 706-718 (2019-12-27)
Extracellular vesicles are endogenous biological nanoparticles that have potential for use as therapeutic nanoparticles or as delivery vehicles for therapeutic agents. Milk nanovesicles (MNV) are extracellular vesicles isolated from bovine milk that have been explored for use as delivery vehicles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service