Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1309057

[KO Validated]Anti-CD44 antibody produced in mouse

new

[Knockout Validated]purified monoclonal antibody, buffered aqueous solution

Synonym(s):

Filamin-A, FLN-A, Actin-binding protein 280, ABP-280, Alpha-filamin, Endothelial actin-binding protein, Filamin-1, Non-muscle filamin, FLNA, FLN, FLN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

(Human)

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

form

liquid

mol wt

280739Da

species reactivity

human, rat, mouse

technique(s)

western blot: 1:4000
western blot: 1:500-1:2000

isotype

IgG1

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... FLNA(2316)

General description

Promotes orthogonal branching of actin filaments and links actin filaments to membrane glycoproteins. Anchors various transmembrane proteins to the actin cytoskeleton and serves as a scaffold for a wide range of cytoplasmic signaling proteins. Interaction with FLNA may allow neuroblast migration from the ventricular zone into the cortical plate. Tethers cell surface-localized furin, modulates its rate of internalization and directs its intracellular trafficking (By similarity). Involved in ciliogenesis.

Specificity

Purified His-tagged FLNA protein was used to produced this monoclonal antibody.

Physical form

Purified monoclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein G column, followed by dialysis against PBS.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service