Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1404390

Sigma-Aldrich

Monoclonal Anti-SNX1 antibody produced in mouse

clone 1E3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

HsT17379, MGC8664, SNX1A, Vps5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.21 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNX1(6642)

General description

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This endosomal protein regulates the cell-surface expression of epidermal growth factor receptor. This protein also has a role in sorting protease-activated receptor-1 from early endosomes to lysosomes. This protein may form oligomeric complexes with family members. This gene results in three transcript variants encoding distinct isoforms. (provided by RefSeq)

Immunogen

SNX1 (NP_003090, 166 a.a. ~ 275 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KVTTQTSLPLFRSKQFAVKRRFSDFLGLYEKLSEKHSQNGFIVPPPPEKSLIGMTKVKVGKEDSSSAEFLEKRRAALERYLQRIVNHPTMLQDPDVREFLEKEELPRAVG

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service