Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4300238

Sigma-Aldrich

Anti-phospho-NFKBIB (pSer23) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-IKBB antibody produced in rabbit, Anti-TRIP9 antibody produced in rabbit, Anti-nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~48 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100

isotype

IgG

immunogen sequence

(L-G-SP-L-G)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer23)

Gene Information

human ... NFKBIB(4793)

Immunogen

Peptide sequence around phosphorylation site of serine 23 (L-G-S(p)-L-G), according to the protein NFKBIB.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Inhibits NF-kappa-B by complexing with and trapping it in the cytoplasm. However, the unphosphorylated form resynthesized after cell stimulation is able to bind NF-kappa-B allowing its transport to the nucleus and protecting it to further IKBA-dependent inactivation. Association with inhibitor kappa B-interacting NKIRAS1 and NKIRAS2 prevent its phosphorylation rendering it more resistant to degradation, explaining its slower degradation.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Sun et al.
Gene, 723, 143986-143986 (2019-07-20)
Diabetic nephropathy (DN) is the leading cause of end-stage renal disease. Accumulating evidence shows that microRNAs play important roles in diabetic kidney. However, the potential role of MicroRNA-544 (miR-544) in DN remains unclear. In this study, we explored the role
Yinan Zhou et al.
Cell biology international, 43(5), 495-505 (2019-02-15)
Peritoneal dialysis (PD) is often used to treat patients with end stage renal disease, and its long-term complications include excessive inflammation and oxidative stress. Allograft inflammatory factor 1 (AIF-1), as a cytoplasmic protein, is originally identified from infiltrating macrophages, and
Gang Liu et al.
Cellular and molecular neurobiology, 37(5), 843-855 (2016-08-31)
FK-506 (Tacrolimus) is a very commonly used immunomodulatory agent that plays important roles in modulating the calcium-dependent phosphoserine-phosphothreonine protein phosphatase calcineurin and thus inhibits calcineurin-mediated secondary neuronal damage. The biological function of FK-506 in the spinal cord has not been
Nan Tang et al.
Microbial pathogenesis, 113, 168-175 (2017-10-24)
In this study, we investigated the anticancer potentials of Rhein, an anthraquinone derivative of most commonly used Chinese rhubarb on the rat F98 glioma cells. The experimental studies revealed that Rhein induced cell cycle arrest, caspase mediated apoptosis. It results
Heyu Zhang et al.
Frontiers in neurology, 9, 931-931 (2018-11-22)
Intracerebral hemorrhage (ICH) has high morbidity and mortality, with no effective treatment at present. One possible therapeutic strategy involves the use of mesenchymal stem cells (MSCs), which have shown promise in experimental models and have great potential for treating nervous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service