Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700909

Sigma-Aldrich

Monoclonal Anti-CD1b antibody produced in mouse

clone SN13

Synonym(s):

R1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SN13, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: 2 μg/mL

accession no.

NM_001764

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB4700809SAB4700031SAB4700867
conjugate

unconjugated

conjugate

unconjugated

conjugate

phycoerythrin (R-PE) conjugate

conjugate

unconjugated

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human, nonhuman primates

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

clone

SN13, monoclonal

clone

CBRM1/5, monoclonal

clone

HI149, monoclonal

clone

ICRF44, monoclonal

Gene Information

human ... CD1B(910)

Gene Information

human ... ITGAM(3684)

Gene Information

human ... CD1A(909)

Gene Information

human ... ITGAM(3684)

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

General description

The mouse monoclonal antibody SN13 (also known as K5-1B8) recognizes CD1b, a 44 kDa type I glycoprotein associated with beta2-microglobulin. It is expressed on dendritic cells, Langerhans cells, thymocytes, and T acute lymphoblastic leukemia cells.

Immunogen

A cell membrane antigen preparation that was isolated from normal human thymocytes

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service