SAB4702022
Monoclonal Anti-CD23 APC produced in mouse

Synonym(s):
BLAST-2, CLEC4J, FCE2, FCER2, Fc-epsilon-RII, IGEBF
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
General description
CD23 (Fc epsilon RII), the low affinity IgE receptor, is a 45 kDa type II membrane glycoprotein expressed more or less on eosinophils, follicular dendritic cells, Langerhans cells, mature B cells (mainly upon activation), EBV-transformed lymphoblasts, monocytes, and subpopulation of platelets. A soluble form of 37 kDa and other its fragments were also described. CD23 mediates IgE-dependent cytotoxicity by eosinophils and macrophages, and downregulates IgE secretion in response to high levels of IgE, involving release of pro-inflammatory cytokines.
Specificity
The mouse monoclonal antibody EBVCS-5 recognizes an extracellular epitope located in the stalk region of human low affinity IgE receptor (CD23) between the 37 and 25 kDa cleavage sites.
Immunogen
EBV-transformed human cells
Physical form
Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide
Preparation Note
Purified antibody is conjugated with activated allophycocyanin (APC) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.
Storage and Stability
Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.
Other Notes
The antibody solution should be gently mixed before use.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service