Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4702023

Monoclonal Anti-CD23 PE-Cyanine7 produced in mouse

new

Synonym(s):

BLAST-2, CLEC4J, FCE2, FCER2, Fc-epsilon-RII, IGEBF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

PE-Cy7 conjugate

Quality Level

antibody product type

primary antibodies

clone

EBVCS-5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1κ

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FCER2(2208)

General description

CD23 (Fc epsilon RII), the low affinity IgE receptor, is a 45 kDa type II membrane glycoprotein expressed more or less on eosinophils, follicular dendritic cells, Langerhans cells, mature B cells (mainly upon activation), EBV-transformed lymphoblasts, monocytes, and subpopulation of platelets. A soluble form of 37 kDa and other its fragments were also described. CD23 mediates IgE-dependent cytotoxicity by eosinophils and macrophages, and downregulates IgE secretion in response to high levels of IgE, involving release of pro-inflammatory cytokines.

Specificity

The mouse monoclonal antibody EBVCS-5 recognizes an extracellular epitope located in the stalk region of human low affinity IgE receptor (CD23) between the 37 and 25 kDa cleavage sites.

Immunogen

EBV-transformed human cells

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with activated tandem dye of R-phycoerythrin-cyanine 7 (PE-Cy7) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service