SAB4702024
Monoclonal Anti-CD23 PerCP-Cyanine5.5 produced in mouse

Synonym(s):
BLAST-2, CLEC4J, FCE2, FCER2, Fc-epsilon-RII, IGEBF
About This Item
conjugate
PerCP-Cy5.5 conjugate
Quality Level
antibody product type
primary antibodies
clone
EBVCS-5, monoclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
flow cytometry: suitable
isotype
IgG1κ
UniProt accession no.
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... FCER2(2208)
General description
Specificity
Immunogen
Physical form
Preparation Note
Storage and Stability
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service