Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5702176

Sigma-Aldrich

Anti-RGAP1 Antibody, clone 10F7U6, Rabbit Monoclonal

Synonym(s):

RACGAP1, CYK4, HsCYK-4, ID-GAP, MgcRacGAP, rac GTPase-activating protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

10F7U6, monoclonal

form

liquid

mol wt

70 kDa

species reactivity

rat, rat, human

concentration

0.50 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MDTMMLNVRNLFEQLVRRVEILSEGNEVQFIQLAKDFEDFRKKWQRTDHELGKYKDLLMKAETERSALDVKLKHARNQVDVEIKRRQRAEADCEKLERQI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RACGAP1(29127)

General description

This gene encodes a GTPase-activating protein (GAP) that is a compoment of the centralspindlin complex. This protein binds activated forms of Rho GTPases and stimulates GTP hydrolysis, which results in negative regulation of Rho-mediated signals. This protein plays a regulatory role in cytokinesis, cell growth, and differentiation. Alternatively spliced transcript variants have been found for this gene. There is a pseudogene for this gene on chromosome 12.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RGAP1 (Q9H0H5).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service