Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5702406

Sigma-Aldrich

Anti-Plexin A1 Antibody, clone 2A5Z6, Rabbit Monoclonal

Synonym(s):

NOV, NOVP, PLEXIN-A1, PLXN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

2A5Z6, monoclonal

form

liquid

mol wt

250 kDa

species reactivity

rat, human, rat

concentration

0.7 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

SVQSCPHGLGSTDNVNKLLLLDYAANRLLACGSASQGICQFLRLDDLFKLGEPHHRKEHYLSSVQEAGSMAGVLIAGPPGQGQAKLFVGTPIDGKSEYFPT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLXNA1(5361)

General description

Coreceptor for SEMA3A, SEMA3C, SEMA3F and SEMA6D. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm (By similarity.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Plexin A1 (Q9UIW2).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service