SAB5702427
Anti-SLC3A2/CD98hc Antibody, clone 1W1U9, Rabbit Monoclonal
Synonym(s):
4F2, 4F2HC, 4T2HC, CD98, CD98HC, MDU1, NACAE
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
biological source
rabbit
Quality Level
material
colorless
clone
1W1U9, monoclonal
form
liquid
mol wt
90 kDa
species reactivity
human
concentration
0.53 mg/mL
technique(s)
western blot: 1:500 - 1:2000
color
colorless
General description
This gene is a member of the solute carrier family and encodes a cell surface, transmembrane protein. The protein exists as the heavy chain of a heterodimer, covalently bound through di-sulfide bonds to one of several possible light chains. The encoded transporter plays a role in regulation of intracellular calcium levels and transports L-type amino acids. Alternatively spliced transcript variants, encoding different isoforms, have been characterized. [provided by RefSeq, Nov 2010]
Immunogen
A synthetic peptide corresponding to a sequence within amino acids 350-450 of human SLC3A2/CD98hc (P08195).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Application
WB
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service