Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5702486

Sigma-Aldrich

[KO Validated]Anti-MLH1 Antibody, clone 8K1T2, Rabbit Monoclonal

Synonym(s):

COCA2, FCC2, HNPCC, HNPCC2, hMLH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

8K1T2, monoclonal

form

liquid

mol wt

85 kDa

species reactivity

mouse, mouse

concentration

0.69 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200
western blot: 1:10000 - 1:68900

color

colorless

isotype

IgG

immunogen sequence

PGLAGPSGEMVKSTTSLTSSSTSGSSDKVYAHQMVRTDSREQKLDAFLQPLSKPLSSQPQAIVTEDKTDISSGRARQQDEEMLELPAPAEVAAKNQSLEGD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MLH1(4292)

General description

This gene was identified as a locus frequently mutated in hereditary nonpolyposis colon cancer (HNPCC). It is a human homolog of the E. coli DNA mismatch repair gene mutL, consistent with the characteristic alterations in microsatellite sequences (RER+phenotype) found in HNPCC. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Additional transcript variants have been described, but their full-length natures have not been determined.[provided by RefSeq, Nov 2009]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 350-450 of human MLH1 (NP_000240.1).
PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service