SAB5702501
Anti-Gephyrin Antibody, clone 9D0B0, Rabbit Monoclonal
Synonym(s):
GEPH, GPH, GPHRYN, HKPX1, MOCODC
About This Item
biological source
rabbit
Quality Level
material
colorless
clone
9D0B0, monoclonal
form
liquid
mol wt
93 kDa
species reactivity
rat, human, rat
concentration
0.4 mg/mL
technique(s)
western blot: 1:500 - 1:2000
color
colorless
isotype
IgG
immunogen sequence
MATEGMILTNHDHQIRVGVLTVSDSCFRNLAEDRSGINLKDLVQDPSLLGGTISAYKIVPDEIEEIKETLIDWCDEKELNLILTTGGTGFAPRDVTPEAT
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... GPHN(10243)
General description
Immunogen
Application
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service