Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB5703002

Sigma-Aldrich

Anti-PLC gamma 2 (PLCG2) Antibody, clone 7I1C9, Rabbit Monoclonal

Synonym(s):

APLAID, FCAS3, PLC-IV, PLC-gamma-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

7I1C9, monoclonal

form

liquid

mol wt

150 kDa

species reactivity

rat, human, rat

concentration

0.512 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

RHFVLGTSAYFESLVELVSYYEKHSLYRKMRLRYPVTPELLERYNMERDINSLYDVSRMYVDPSEINPSMPQRTVKALYDYKAKRSDELSFCRGALIHNVS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLCG2(5336)

General description

The protein encoded by this gene is a transmembrane signaling enzyme that catalyzes the conversion of 1-phosphatidyl-1D-myo-inositol 4,5-bisphosphate to 1D-myo-inositol 1,4,5-trisphosphate (IP3) and diacylglycerol (DAG) using calcium as a cofactor. IP3 and DAG are second messenger molecules important for transmitting signals from growth factor receptors and immune system receptors across the cell membrane. Mutations in this gene have been found in autoinflammation, antibody deficiency, and immune dysregulation syndrome and familial cold autoinflammatory syndrome 3. [provided by RefSeq, Mar 2014]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 700-800 of human PLC gamma 2 (PLC gamma 2 (PLCG2)) (P16885).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service