Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5703050

Sigma-Aldrich

Anti-Phospho-VANGL2(pSer14/Ser17) Antibody, clone 4C7Y1, Rabbit Monoclonal

Synonym(s):

VANGL2,LPP1,LTAP,STB1,STBM,STBM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

material

colorless

clone

4C7Y1, monoclonal

form

liquid

mol wt

60 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VANGL2(57216)

General description

The protein encoded by this gene is a membrane protein involved in the regulation of planar cell polarity, especially in the stereociliary bundles of the cochlea. The encoded protein transmits directional signals to individual cells or groups of cells in epithelial sheets. This protein is also involved in the development of the neural plate.

Immunogen

A phospho specific peptide corresponding to residues surrounding S14/S17 of human VANGL2
PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.

Application

WB

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service