Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB5900209

Sigma-Aldrich

Alpaca Anti-PCSK9, recombinant single domain Antibody

Recombinant Alpaca(VHH) Nano antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

alpaca

Quality Level

recombinant

expressed in E. coli

conjugate

unconjugated

antibody form

affinity purified VHH antibody

clone

LDL7, monoclonal

assay

>95% (SDS-PAGE)

form

lyophilized solid

mol wt

14.4 kDa
~15-18 kDa (SDS PAGE under reducing condition)

purified by

immobilized metal affinity chromatography (IMAC)

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: 1:3000-1:10000
immunofluorescence: 1:200-1:600
immunohistochemistry: 1:200-1:600

color

colorless

pH

( in PBS, pH 7.4)

isotype

VHH

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PCSK9(255738)

General description

Proprotein convertase subtilisin/kexin type 9 (PCSK9), also known as NARC1 (neural apoptosis regulated convertase), which is a newly identified human secretory subtilase belonging to the proteinase K subfamily of the secretory subtilase family. PCSK9 protein is an enzyme which in humans is encoded by the PCSK9 gene with orthologs found across many species. It is expressed in neuroepithelioma, colon carcinoma, hepatic and pancreatic cell lines, and in Schwann cells. PCSK9 protein is highly expressed in the liver and regulates low density lipoprotein receptor (LDLR) protein levels. Inhibition of PCSK9 protein function is currently being explored as a means of lowering cholesterol levels. Thereby, PCSK9 protein is regarded as a new strategy to treat hypercholesterolemia. PCSK9 protein contributes to cholesterol homeostasis and may have a role in the differentiation of cortical neurons.Alpaca derived anti-PCSK9 VHH single domain antibody (molecular weight: 14.4 kDa), expressed in E. coli under conditions free from animal derived components.It has a 6His tag at its C-terminal.
Single-domain antibody (sdAb), also called nano-antibody, is the variable region isolated from the antigen-binding variable domain (VHH) of camelid heavy-chain-only antibody. It lacks light chains and a first constant domain(CH1). This configuration confers a number of unique advantages over conventional monoclonal antibodies. The small size of just 15 kDa (2~4 nm), single-domain antibody is the smallest antigen-binding fragment that can easily penetrate tissues, allowing them to bind epitopes that are inacessible to conventional antibodies. These single-domain antibodies offer superior chemical and thermal stability and increased solubility than conventional antibodies. All our antibodies are tested to ensure the highest level of specificity, sensitivity and reproducibility.

Specificity

Human Proprotein Convertase Subtilisin/Kexin Type 9 (PCSK9)

Immunogen

Recombinant human PCSK9 (Gln31-Gln692) protein

Application

ELISA, IHC, IF

Packaging

50 µg per vial / 100 µg per vial

Physical form

Affinity purified VHH single domain antibody lyophilized from PBS, pH7.12

Reconstitution

Reconstitute the lyophilized antibody with sterile deionized water or with 50% glycerol (ACS grade or better) to a stock solution of 1 mg/mL. Solubilize for 30 to 60 minutes at room temperature with occasional gentle mixing. Avoid vigorous shaking or vortexing.

Storage and Stability

Stable for 1 year at 4°C from date of receipt.

Other Notes

Storage at 4°C; Do not freeze.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service