SBR00119
Sarecycline Hydrochloride
Synonym(s):
P-005672 HCl, P005672 HCl
About This Item
Recommended Products
storage condition
under inert gas
Quality Level
antibiotic activity spectrum
Gram-negative bacteria
Gram-positive bacteria
mode of action
protein synthesis | inhibits
shipped in
ambient
storage temp.
2-8°C
SMILES string
CN(C)[C@H]1[C@H]2C[C@H]3CC4=C(C=CC(=C4C(=C3C(=O)[C@@]2(C(=C(C1=O)C(=O)N)O)O)O)O)CN(C)OC.Cl
InChI
1S/C24H29N3O8.ClH/c1-26(2)18-13-8-11-7-12-10(9-27(3)35-4)5-6-14(28)16(12)19(29)15(11)21(31)24(13,34)22(32)17(20(18)30)23(25)33;/h5-6,11,13,18,28-29,32,34H,7-9H2,1-4H3,(H2,25,33);1H/t11-,13-,18-,24-;/m0./s1
InChI key
JGPBDCKZLBSHOI-FIPJBXKNSA-N
General description
Biochem/physiol Actions
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service