Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2890

Sigma-Aldrich

BAY-899

≥98% (HPLC)

Synonym(s):

(5S)-N-[2-(4-Fluorophenoxy)pyrimidin-5-yl]-5-(4-fluorophenyl)-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxamide, (S)-N-(2-(4-Fluorophenoxy)pyrimidin-5-yl)-5-(4-fluorophenyl)-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H19F2N5O2
CAS Number:
Molecular Weight:
459.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 165 to 195°, c = 1 in chloroform-d

color

white to beige

solubility

DMSO: 1 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N1[C@@H](C2=CC=C(F)C=C2)C3=C(N=CC=C3)CC1)NC4=CN=C(OC5=CC=C(F)C=C5)N=C4

InChI

1S/C25H19F2N5O2/c26-17-5-3-16(4-6-17)23-21-2-1-12-28-22(21)11-13-32(23)25(33)31-19-14-29-24(30-15-19)34-20-9-7-18(27)8-10-20/h1-10,12,14-15,23H,11,13H2,(H,31,33)/t23-/m0/s1

InChI key

VKQBTIMLSDGNLG-QHCPKHFHSA-N

Biochem/physiol Actions

BAY-899 is an orally available, potent and selective antagonist of the human luteinizing hormone receptor (hLH-R) that reduces sex hormone levels in several animal models. BAY-899 likely binds to a putative rhodopsin-like binding cavity located within the 7-transmembrane region of the hLH-receptor.
orally available, potent and selective antagonist of the human luteinizing hormone receptor (hLH-R)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service