Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4008

Sigma-Aldrich

Zuclopenthixol

≥98% (HPLC)

Synonym(s):

(Z)-4-[3-(2-chloro-9H-thioxanthen-9-ylidene)propyl]-1-piperazineethanol, 4-[(3Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)propyl]-1-piperazineethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25ClN2OS
CAS Number:
Molecular Weight:
400.96
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

S1c2c(cc(cc2)Cl)\C(=C/CCN4CCN(CC4)CCO)\c3c1cccc3

InChI key

WFPIAZLQTJBIFN-DVZOWYKESA-N

Biochem/physiol Actions

Oral and potent antagonist of D1/D2 dopamine receptors, α1-adrenergic receptors and 5-HT2 receptors.



Zuclopenthixol, a typical antipsychotic, is an oral and potent antagonist of D1/D2 dopamine receptors, α1-adrenergic receptors and 5-HT2 receptors. Zuclopenthixol is approved in some countries for treatment of schizophrenia and other psychoses. Zuclopenthixol is a cis isomer of neuroleptic clopenthixol.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service