Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2002

Sigma-Aldrich

TFIIA-p12 human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

HsT18745, TF2A2, TFIIA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

biological source

human

recombinant

expressed in E. coli

assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~14.5 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

300 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... GTF2A2(2958)

Biochem/physiol Actions

p12 is a small subunit ( γ ) of the transcription factor IIA and has been shown to be required for both basal and activated transcription. Recombinant p12, along with two other subunits ( α and β ) can potentiate transcriptional activation, whereas p12 along with β subunit is able to function in an anti-repression for reconstitued TFIIA, containing all three subunits.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Sun et al.
Genes & development, 8(19), 2336-2348 (1994-10-01)
Human TFIIA activity is composed of three subunits (alpha, beta, gamma). Here we report the isolation of a human cDNA clone encoding the gamma-subunit and the reconstitution of TFIIA activity from recombinant polypeptides (holo-TFIIA). Protein-protein interaction analysis established that the
J DeJong et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(8), 3313-3317 (1995-04-11)
The human general transcription factor TFIIA is one of several factors involved in specific transcription by RNA polymerase II, possibly by regulating the activity of the TATA-binding subunit (TBP) of TFIID. TFIIA purified from HeLa extracts consists of 35-, 19-
J Ozer et al.
Genes & development, 8(19), 2324-2335 (1994-10-01)
TFIIA is thought to play an important role in transcriptional regulation in higher eukaryotes, but its precise function is unclear. A human cDNA encoding a protein with 45% identity to the small subunit of yeast TFIIA has been isolated. TFIIA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service