Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5261

Sigma-Aldrich

SETDB2, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CLLD8, CLLL8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~91 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SETDB2(83852)

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A functional AT/G polymorphism in the 5'-untranslated region of SETDB2 in the IgE locus on human chromosome 13q14
R J Holt
Genes and Immunity (2015)
Left-Right Axis Differentiation and Functional Lateralization: a Haplotype in the Methyltransferase Encoding Gene SETDB2 Might Mediate Handedness in Healthy Adults.
Ocklenburg s
Molecular Neurobiology (2016)
CLLD8/KMT1F is a lysine methyltransferase that is important for chromosome segregation.
Falandry C
The Journal of Biological Chemistry (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service