Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5263

Sigma-Aldrich

KAT4, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

BA2R, CCG1, CCGS, DYT3, DYT3/TAF1, N-TAF1, NSCL2, OF, P250, TAF1, TAF2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~155 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TAF1(6872)

General description

KAT4 is a DNA-binding protein complex required for RNA polymerase II-mediated transcription of many, if not all, protein-encoding genes in eukaryotic cells. KAT4 plays a key role in the initiation process, since it binds to the TATA element to form a complex that nucleates the assembly of the other components into a preinitiation complex and that may be stable through multiple rounds of transcription. KAT4 may be targeted to specific chromatin-bound promoters and may play a key role in chromatin recognition. KAT4 also serves to link the control of transcription to the cell cycle.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service