US115251
10 Position Slide Carrier for MIDAS®

For use with MIDAS® III-Plus Automated Stainer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
1 ea of
compatibility
for use with Midas® III-Plus Automated Stainer
shipped in
ambient
Legal Information
MIDAS is a registered trademark of Merck KGaA, Darmstadt, Germany
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service