Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB1198

Sigma-Aldrich

Anti-TSTA3 Antibody, clone 2N10-H1 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2N10-H1, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~35.89 kDa
observed mol wt ~38 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2N10-H1 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects human GDP-L-fucose synthase. It targets an epitope within 20 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human GDP-L-fucose synthase.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected TSTA3 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected TSTA3 in MCF-7 cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100-1,000 dilution from a representative lot detected TSTA3 in human testis and human kidney tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TSTA3 in HepG2 cells.

Affinity Binding Assay: A representative lot of this antibody bound TSTA3 peptide with a KD of 1.7 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-TSTA3, clone 2N10-H1 ZooMAb®, Cat. No. ZRB1198, is a recombinant Rabbit monoclonal antibody that specifically targets TSTA3 and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry, and Western Blotting.

Target description

GDP-L-fucose synthase (UniProt: Q13630; also known as EC:1.1.1.271, GDP-4-keto-6-deoxy-D-mannose-3,5-epimerase-4-reductase, Protein FX, Red cell NADP(H)-binding protein, Short-chain dehydrogenase/reductase family 4E member 1) is encoded by the GFUS (also known as SDR4E1, TSTA3) gene (Gene ID: 7264) in human. In mammals, fucosylated glycans play important roles in ABO blood group antigens, host-microbe interactions, and several human diseases, including rheumatoid arthritis, pancreatic cancer, and hepatocellular carcinoma. GDP-L-fucose synthase is a homodimeric protein of the NAD(P)-dependent epimerase/dehydratase family that is responsible for the last step of the major metabolic pathway resulting in GDP-L-fucose synthesis from GDP-D-mannose. It catalyzes a combined epimerase and NADPH-dependent reductase reaction, converting GDP-4-keto-6-D-deoxymannose to GDP-L-fucose. This reaction involves epimerization at both C-3 and C-5 followed by an NADPH-dependent reduction of the carbonyl at C-4. GDP-L-fucose synthase is also reported to act as the main autoantigen for a brain-infiltrating T cell clone. It is recognized by cerebrospinal fluid-infiltrating CD4+ T cells from HLA-DRB3*-positive subjects. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Santos, R., et al. (2018). Sci. Trans. Med. 10(462); eaat4301; Zhou, H., et al. (2013). Acta Biochim. Biophys. Sinica. 45(9); 720-725).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service