Skip to Content
MilliporeSigma
All Photos(18)

Key Documents

ZRB1551

Sigma-Aldrich

Anti-HDAC7 Antibody, clone 1L17 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Histone deacetylase 7;EC:3.5.1.98;HD7;Histone deacetylase 7A;HD7a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1L17, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 102.93 kDa
observed mol wt ~110 kDa

purified by

using Protein A

species reactivity

human, rat, mouse

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1L17 is a Rabbit recombinant monoclonal antibody that specifically detects Histone Deacetylase 7 (HDAC7). It targets an epitope within 21 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the N-terminal region of human Histone Deacetylase 7 (HDAC7).

Application

Quality Control Testing

Evaluated by Western Blotting in Rat Thymus tissue lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected HDAC7 in Rat Thymus tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected HDAC7 in HeLa and NIH3T3 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected HDAC7 in human placenta tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected HDAC7 in A431, HeLa, and NIH 3T3 cells.

Affinity Binding Assay: A representative lot of this antibody bound HDAC7 with a KD of 1.0 x 10-12 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Histone deacetylase 7 (UniProt: Q8WUI4; also known as EC:3.5.1.98, HD7, Histone deacetylase 7A, HD7a) is encoded by the HDAC7 (also known as HDAC7A) gene (Gene ID: 51564) in human. Histone acetylation and deacetylation play a major role in determining chromatin structure and function. These modifications are achieved by histone acetyltransferases (HATs) and histone deacetylases (HDACs). In the deacetylated state, specific basic amino acids in histones are positively charged and interact with DNA s negatively charged phosphate groups and with negatively charged patches on neighboring nucleosomes. This promotes chromatin condensation to form heterochromatin. On the other hand, acetylation neutralizes these positive charges and promoting the less condensed and more accessible chromatin conformation known as euchromatin. HDAC7 is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). It shuttles between the nucleus and the cytoplasm and in the nucleus, it associates with distinct subnuclear dot-like structures. Its nuclear export signal is localized in amino acids 918-952. Its deacetylase activity maps to a C-terminal domain and is dependent on its interaction with other class I HDACs in the nucleus. HDAC7 can undergo phosphorylation by PKN1 and PKN2 that impairs its nuclear import. Its phosphorylation on serine 155 by MARK2, MARK3, and PRKD1 is shown to promote its interaction with 14-3-3 proteins and facilitate its export from the nucleus. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service