Skip to Content
MilliporeSigma
All Photos(11)

Key Documents

ZRB2036

Sigma-Aldrich

Anti-PBR/TSPO Antibody, clone 1E8 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Mitochondrial benzodiazepine receptor, PKBS, Peripheral-type benzodiazepine receptor, Translocator protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E8, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 18.83 kDa
observed mol wt ~16 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal cytoplasmic domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... TSPO(706)

Related Categories

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E8 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Translocator protein (PBR/TSPO). It targets an epitope within 14 amino acids from the C-terminal, third cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the third cytoplasmic domain within the C-terminal region of human Translocator protein (PBR/TSPO).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PBR/TSPO in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PBR/TSPO in U87-MG, and Human kidney tissue lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected PBR/TSPO in Human colon tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PBR/TSPO in A431 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Translocator protein (UniProt: P30536; also known as Mitochondrial benzodiazepine receptor, PKBS, Peripheral-type benzodiazepine receptor, PBR) is encoded by the TSPO (also known as BZRP, MBR) gene (Gene ID: 706) in human. TSPO is a multi-pass outer mitochondrial membrane protein with three mitochondrial intermembrane domain, five transmembrane domains, and three cytoplasmic domains. It is expressed in many tissues with highest expression observed in tissues that synthesize steroids. It plays an important role in translocating cholesterol from the cytoplasm into mitochondria, which is a rate limiting step in steroid synthesis. TSPO expression is also observed in the microglia in the central nervous system, and its expression is greatly increased upon microglial activation. It is considered as a sensitive biomarker of brain damage and neurodegeneration, particularly of inflammation and reactive gliosis. TSPO is shown to bind with voltage-dependent anion channel (VDAC) and the adenine nucleotide transporter (ANT).  It can bind protoporphyrin IX and may play a role in the transport of porphyrins and heme. It promotes the transport of cholesterol across mitochondrial membranes and may play a role in lipid metabolism. However, it is apparently not required for steroid hormone biosynthesis.  Two isoforms of TSPO are descried that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Yao, R., et al. (2020). Front. Pharmacol. 11; 986; Rupprecht, R., et al. (2010). Nat. Rev. Drug Dis. 9; 971-988; Papadopoulos, V., et al. (2006). Trends Pharmacol. Sci. 27 (8); 402-409). 

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service