Skip to Content
MilliporeSigma
All Photos(15)

Key Documents

ZRB2037

Sigma-Aldrich

Anti-MEF2C Antibody, clone 1D21 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Myocyte-specific enhancer factor 2C;Myocyte enhancer factor 2C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D21, recombinant monoclonal

description

1D21 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 51.22 kDa
observed mol wt ~34 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MEF2C(4208)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1D21 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Myocyte enhancer factor 2C (MEF2C). It targets an epitope within 17 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal half of human Myocyte enhancer factor 2C (MEF2C).

Application

Quality Control Testing

Evaluated by Western Blotting with His-tagged recombinant fragment of Human MEF2C.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected His-tagged recombinant fragment of Human MEF2C.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:100-1,000 dilution from a representative lot detected MEF2C in human cerebral cortex and human tonsil tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected MEF2C in HeLa cells.

Affinity Binding Assay:: A representative lot of this antibody bound MEF2C peptide with a KD of 1.7 x 10-7 in an Affinity Binding Assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Myocyte-specific enhancer factor 2C (UniProt: Q06413; also known as Myocyte enhancer factor 2C) is encoded by the MEF2C gene (Gene ID: 4208) in human. Myocyte enhancer factor 2 (MEF2) family of transcription factors in vertebrates consists of four members: MEF2A, MEF2B, MEF2C, and MEF2D. They are highly conserved and are critical for several functions in a variety of different cell types, including cell differentiation and extracellular stimuli response. MEF2C is expressed in brain and skeletal muscle and its expression is highest during the early stages of postnatal development. It binds specifically to the MEF2 element present in the regulatory regions of many muscle-specific genes. MEF2C is reported to be involved in cardiac morphogenesis and myogenesis, and in vascular development. It is also required for proper development of megakaryocytes and platelets and for bone marrow B-lymphopoiesis. It is shown to be crucial for normal neuronal development, distribution, and electrical activity in the neocortex and plays a role in hippocampal-dependent learning and memory by suppressing the number of excitatory synapses and thus regulating basal and evoked synaptic transmission. Six isoforms of MF2C have been described that are produced by alternative splicing. Isoforms that lack the repressor domain (aa 368-399) are more active. Its β-domain (aa 271-278) that is required for enhancement of transcriptional activity is missing in a number of isoforms. Phosphorylation of MEF2C on serine 59 enhances its DNA binding activity and phosphorylation on serine 396 is required for sumoylation on lysine 391 and inhibition of its transcriptional activity. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Chaudhary, R., et al. (2021). Heliyon. 7(4); e06854; Tang, C-M., et al. (2016). Sci. Rep. 6; Article 36146; Kang, J., et al. (2006). BMC Biochem. 7; 5).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service