Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

ZRB2317

Anti-PIK3R3 Antibody, clone 1F2 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

PI3-kinase regulatory subunit gamma, PI3-kinase subunit p55-gamma, PI3K regulatory subunit gamma, Phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma, Phosphatidylinositol 3-kinase regulatory subunit gamma, PtdIns-3-kinase regulatory subunit gamma, PtdIns-3-kinase regulatory subunit p55-gamma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 54.45 kDa
observed mol wt ~58 kDa

purified by

using Protein A

species reactivity

human, rat, mouse

species reactivity (predicted by homology)

monkey, nonhuman primates

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... PIK3R3(8503)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1F2 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects PIK3R3. It targets an epitope within 24 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the C-terminal half of human PIK3R3.

Application

Quality Control Testing

Evaluated by Western Blotting in L6 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected PIK3R3 in L6 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected PIK3R3 in lysates from Neuro-2a and MDA-MB-435 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected PIK3R3 in human cerebral cortex tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound with PIK3R3 peptide with at least ten thousand-fold (10,000X) higher affinity than with non-specific control peptide.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PIK3R3 in SH-SY5Y cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Phosphatidylinositol 3-kinase regulatory subunit gamma (UniProt: Q92569; also known as PI3-kinase regulatory subunit gamma, PtdIns-3-kinase regulatory subunit gamma, Phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma, PI3-kinase subunit p55-gamma, PtdIns-3-kinase regulatory subunit p55-gamma) is encoded by the PIK3R3 gene (Gene ID: 8503) in human. PI3K phosphorylates phosphatidylinositol and similar compounds, which then serve as second messengers in growth signaling pathways. PI3K is composed of a catalytic and a regulatory subunit. The protein encoded by this gene represents a regulatory subunit of PI3K. The encoded protein contains two SH2 domains (SH2 1 and SH2 2) through which it binds activated protein tyrosine kinases to regulate their kinase activity. During insulin stimulation, it also binds to IRS-1. PI3K is expressed in various tissues, with notable high levels in brain and testis. It is also found in lower levels in adipose tissue, kidney, heart, lung and skeletal muscle. It is predominantly localized in the cytoplasm and can translocate to the plasma membrane upon activation. Three isoforms of PI3K have been described that are produced by alternative splicing. Dysregulation of the PI3K signaling pathway, often due to mutations in PIK3R3 or its associated components, can lead to uncontrolled cell growth and survival, contributing to tumorigenesis. It is indicated that PIK3R3 promotes colorectal cancer metastasis by positively regulating cell migration and invasion. PIK3R3 also plays a pivotal role in ovarian cancer development and its expression is upregulated in ovarian cancer as well as ovarian cancer stem cells. Inhibition of PIK3R3 reduced the spheroid size of ovarian cancer stem cells. PIK3R3 is suggested to be a useful prognostic marker for ovarian cancer and can therefore be exploited for developing therapeutic strategies for this cancer type. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Sohn, E.J., et al. (2022). BMC Cancer. 22(1):708; Wang, G., et al. (2014). Mol Cancer Ther. 13(7);1837-1847).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service