Skip to Content
MilliporeSigma
All Photos(10)

Key Documents

ZRB2780

Sigma-Aldrich

Anti-HLA-DRB1 Antibody, clone 4D12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

HLA class II histocompatibility antigen, DRB1 beta chain, Human leukocyte antigen DRB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4D12, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 30 kDa
observed mol wt ~32 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... HLA-DRB1(3123)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 4D12 is a ZooMAb® rabbit recombinant monoclonal antibody that detects Human leukocyte antigen DRB1 (HLA-DRB1). It targets an epitope within 15 amino acids from the N-terminal extracellular domain of the protein.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal extracellular domain of human HLA-DRB1.

Application

Quality Control Testing

Evaluated by Western Blotting in Ramos cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HLA-DRB1 in Ramos cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected HLA-DRB1 in lysates from human PBMCs and human tonsil tissue.

Affinity Binding Assay:: The binding affinity of this antibody to HLA-DRB1 peptide was at least one thousand-fold (1,000X) higher than that obtained with the non-specific control peptide.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected HLA-DRB1 in Raji and Ramos cells.

Flow Cytometry Analysis: 1 µg from a representative lot detected HLA-DRB1 in human PBMC.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected HLA-DRB1 in human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

HLA class II histocompatibility antigen, DRB1 beta chain (UniProt: P01911; also known as Human leukocyte antigen DRB1) is encoded by the HLA-DRB1 gene (Gene ID: 3123) in human. HLA-DRB1 is a protein involved in the immune system′s recognition and presentation of foreign antigens to T cells. It belongs to the major histocompatibility complex (MHC) class II proteins. It consists of an alpha chain and a beta chain. The beta chain, encoded by the HLA-DRB1 gene, contains extracellular domains responsible for antigen binding and presentation to T cells. The primary function of HLA-DRB1 is to present antigenic peptides to CD4+ T cells, facilitating immune responses against pathogens. It plays a crucial role in adaptive immunity by presenting antigens derived from external sources, such as viruses or bacteria, to T lymphocytes. HLA-DRB1 is expressed on antigen-presenting cells (APCs) like dendritic cells, macrophages, and B cells. Its expression is regulated by various factors, including cytokines and immune stimulation. In different tissues, HLA-DRB1 expression can vary, indicating tissue-specific roles in immune responses. It forms complexes with other MHC class II molecules, chaperones, and accessory proteins in the endosomal-lysosomal pathway to facilitate antigen processing and presentation. HLA-DRB1 is associated with several autoimmune diseases, including rheumatoid arthritis and certain forms of juvenile idiopathic arthritis. Specific HLA-DRB1 alleles confer susceptibility or protection to these diseases and contribute to the pathogenesis by influencing antigen presentation and immune responses. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service