SPR-480B
StressMarq Biosciences Inc
StressMarq Tau-441 (2N4R) Wild-Type Pre-formed Fibrils
100 µg
Synonym(s):
Tau PFFs, MAPT, Truncated Tau Protein Aggregate, Paired Helical Filament-Tau, G Protein Beta1/Gamma2 Subunit-Interacting Factor 1, Isoform 2, tubulin-associated unit, 95-amino acid Tau protein fragment
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Molecular Weight:
45.85
Recommended Products
assay
95
concentration
2 mg/mL
application(s)
life science and biopharma
biological source
human
recombinant
E. coli
mol wt
predicted mol wt 45.85 kDa
pH
7.4
storage temp.
-80 °C
storage condition
avoid repeated freeze/thaw cycles
manufacturer/tradename
StressMarq Biosciences Inc
Related Categories
General description
Tau-441 (2N4R) Wild-Type Pre-formed Fibrils are expressed in E. coli and Ion-exchange Purified. Certified >95% pure using SDS-PAGE analysis. Low endotoxin <5 EU/mL @ 2mg/mL. Validated for WB | SDS-PAGE | In vivo assay | In vitro assay. Monomer source is catalog# SPR-479.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.