Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL10018

Cannabigerovarinic acid

phyproof® Reference Substance

Synonym(s):

CBGVA, Cannabigerovarinic acid A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O4
CAS Number:
Molecular Weight:
332.43
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages
pharmaceutical

storage temp.

2-8°C

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

This substance is tested for its content of d8-THC, d9-THC, and d9-THCA. Results are given on each COA. Acceptable maximum limits were set to 0.10 % each, and to 0.20 % for the sum of d9-THC and d9-THCA (in analogy to the cannabidiol monograph in DAC).

Other Notes

This compound is commonly found in plants of the genus: cannabis

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service