Key Documents
RPC0005
TriForest
TriForest Autoclavable Polycarbonate Serum Vial
5mL, 1600 per case
Synonym(s):
Autoclavable Bottles, Polycarbonate Bottles, Media Bottles Serum Vial
About This Item
PDF Icon
Shipped & Sold By TriForest
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
TriForest Enterprises
General description
TriForest polycarbonate (PC) vial is a safer alternative to glass vial. These vials are ideal for packaging serum, veterinary, pharmaceuticals, cosmeceutical, nutraceutical extracts, as well as sampling, storage, and production of steam sterilized biologicals.
  • Made with virgin polycarbonate resin, durable, chemically inert, non-leaching, optically clear
  • Molded seal ring inside cap fits tightly against bottle neck
  • No fluid entrapment and back flow contamination
  • Ideal for steam sterilization
  • Shatterproof, durable, reusable
  • Manufactured in a Class 10K cleanroom
  • Sold by the case - 1600 bottles per case
Choose from one of the most recent versions:
Certificates of Analysis (COA)