Key Documents
500092
WPI
WPI Adson Forceps
12 cm, Straight, 1x2 Teeth, Stainless Steel
About This Item
PDF Icon
A Trusted Marketplace Seller
PDF Icon
Have a question?
General description
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Choose from one of the most recent versions:
Certificates of Analysis (COA)