Key Documents
500355
WPI
WPI Disposable Scalpel
#23, 10/Box, Sterile
About This Item
PDF Icon
A Trusted Marketplace Seller
PDF Icon
Have a question?
General description
The surgical stainless steel blades are installed on plastic handles, and each unit comes sterile and individually wrapped. These are perfect for one time surgical uses, dissections, crafts, student use, woodworking, taxidermy, clay modeling and more.
Each disposable knife comes with a cap to protect the blade for safe disposal or to save the used scalpel for another application.
The plastic handle of the scalpel is textured to ensure a firm grip. It is lightweight for comfortable use. And the handle includes a handy, integrated 6 cm metric ruler.
Choose from one of the most recent versions:
Certificates of Analysis (COA)