Key Documents
502041
WPI
WPI Cover Slips
Diameter 8 mm, Pkg. 100
About This Item
PDF Icon
A Trusted Marketplace Seller
PDF Icon
Have a question?
General description
These cover slips (made of German glass) can be used for growing and culturing cells that normally have poor adhesion to plastic surfaces. They are small enough to be placed in the micro plate or other cell culture devices. The 5 mm size will fit inside the 96-well culture plate and leave enough room to pick it up from the bottom of the well with forceps. The 8mm size fits inside the 24-well plates.
Choose from one of the most recent versions:
Certificates of Analysis (COA)