Skip to Content
MilliporeSigma
o-Iodohippuric acid United States Pharmacopeia (USP) Reference Standard

o-Iodohippuric acid

o-Iodohippuric acid United States Pharmacopeia (USP) Reference Standard
Synonym(s):
2-Iodohippuric acid, N-(2-Iodobenzoyl)glycine
Linear Formula:
IC6H4CONHCH2CO2H
CAS Number:
Molecular Weight:
305.07

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing