1344305
USP
o-Iodohippuric acid
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
2-Iodohippuric acid, N-(2-Iodobenzoyl)glycine
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
iodohippuric acid
manufacturer/tradename
USP
mp
170-172 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
OC(=O)CNC(=O)c1ccccc1I
InChI
1S/C9H8INO3/c10-7-4-2-1-3-6(7)9(14)11-5-8(12)13/h1-4H,5H2,(H,11,14)(H,12,13)
InChI key
CORFWQGVBFFZHF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service